missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Prostatic Acid Phosphatase/ACPP DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
Pack of 5
Description
The Human Prostatic Acid Phosphatase/ACPP DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Prostatic Acid Phosphatase/ACPP.
Specifications
Specifications
| Assay Range | 62.50 to 4000 pg/mL |
| Product Type | Solid Phase Sandwich ELISA |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 55 |
| Gene Symbol | ACPP |
| Host Species | Human |
| Quantity | 1 Kit (for 5 Plates) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | acid phosphatase, prostate, ACP3, ACP-3, ACPP, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase |
| Target | Prostatic Acid Phosphatase/ACPP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction