missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Prostatic Acid Phosphatase/ACPP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6240-SP
This item is not returnable.
View return policy
Description
Prostatic Acid Phosphatase/ACPP Monoclonal specifically detects Prostatic Acid Phosphatase/ACPP in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Prostatic Acid Phosphatase/ACPP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P15309 | |
| ACPP | |
| Mouse myeloma cell line NS0-derived recombinant mouse Prostatic Acid Phosphatase/ACPP Lys33-Ser379 Accession # P15309 | |
| 25 μg | |
| Primary | |
| Detects human Prostatic Acid Phosphatase/ACPP in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human TRACP/PAP/ACP5 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 690017 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| acid phosphatase, prostate, ACP3, ACP-3, ACPP, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 55 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction