missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Prostate Tissue Lysate (Adult Normal)

Product Code. 18122178 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18122178 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18122178 Supplier Novus Biologicals™ Supplier No. NBP247057
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Vials of lysate containing 0.1mg of normal or prostatic lysate that has not been subjected to denaturing or reducing conditions. Lysates are guaranteed for 6 months from date of receipt.

These lysates have not been subjected to denaturing or reducing conditions. This allows the tissue or cell lysate to be used in a variety of applications; to study protein-protein interaction, ligand binding, ELISA, immunoprecipitation, 1D and 2D gel electrophoresis, and Western blotting for the detection of specific protein targets. For use in 1D and 2D gel electrophoresis, the addition of a denaturing gel loading buffer with reducing agents may be required. Buffer requirements for performing protein-protein interaction and ligand binding studies can vary significantly from RIPA buffer and may require modifications. In most cases, tissue lysates in RIPA buffer can be used, directly in standard ELISA and immunoprecipitation assays. These lysates are proteomic discovery tools. Researchers should validate and optimize for individual use.

The Human Prostate Tissue Lysate (Adult) is derived from human.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protein-Protein Interaction, Ligand Binding, ELISA, Immunoprecipitation, 1 D and 2 D Gel Electrophoresis, Western Blot
Tissue Prostate
Quantity 0.1 mg
Host Species Human
Content And Storage Store at -80 C. Avoid freeze-thaw cycles.
Target Prostate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.