missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Proprotein Convertase 9/PCSK9 (Research Grade Alirocumab Biosimilar) Alexa Fluor 488-conjugated Antibody, R&D Systems™

Human Monoclonal Antibody
Brand: R&D Systems IC10347G-100UG
This item is not returnable.
View return policy
Description
Proprotein Convertase 9/PCSK9 Monoclonal specifically detects Proprotein Convertase 9/PCSK9 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Proprotein Convertase 9/PCSK9 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 255738 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| Hu135 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 | |
| Human Proprotein Convertase 9/PCSK9 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction