missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Proprotein Convertase 9/PCSK9 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB38883-100
This item is not returnable.
View return policy
Description
Proprotein Convertase 9/PCSK9 Monoclonal specifically detects Proprotein Convertase 9/PCSK9 in Human samples. It is validated for ELISA.
Specifications
| ProproteinConvertase9/PCSK9 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 | |
| Mouse myeloma cell line NS0-derived recombinant human Proprotein Convertase 9/PCSK9 Arg29-Gln692 Accession # Q8NBP7 | |
| 100 μg | |
| 255738 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 499920R | |
| ELISA | |
| Q8NBP7 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human Proprotein Convertase 9/PCSK9 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction