missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Properdin ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EH383RBX10
Additional Details : Weight : 2.50000kg
Description
This gene encodes a plasma glycoprotein that positively regulates the alternative complement pathway of the innate immune system. This protein binds to many microbial surfaces and apoptotic cells and stabilizes the C3- and C5-convertase enzyme complexes in a feedback loop that ultimately leads to formation of the membrane attack complex and lysis of the target cell. Mutations in this gene result in two forms of properdin deficiency, which results in high susceptibility to meningococcal infections.Specifications
P27918 | |
42 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
CFP | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
42 to 10,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
5199 | |
<12% | |
Pre-coated 96 well plate|Standard|Assay Diluent concentrate|Biotinylated Detection Antibody|SAV-HRP|Wash Buffer|Chromogen|Stop Solution|Adhesive Plate Covers | |
10 x 96 Tests | |
Supernatant, 50 μL|Serum, 50 μL|Plasma, 50 μL | |
Properdin | |
4 hr 45 min |