missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Properdin ELISA Kit
Human Properdin quantitates human Properdin in serum, plasma, supernatant.
£540.00 - £2320.00
Specifications
Accession Number | BCFG; Bf; BFD; C3 Proaccelerator; C3/C5 convertase; cfb; Cfp; Complement factor P; complement factor properdin; GBG; H2-Bf; PBF2; PFC; PFD; PROPERDIN; properdin factor, complement; Properdin P factor, complement |
---|---|
Assay Range | 42 to 10,000 pg/mL |
Assay Sensitivity | 42 pg/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17175159
|
Invitrogen™
EH383RBX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
17164683
|
Invitrogen™
EH383RB |
96 Tests |
£540.00
Pack of 96 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
Description
Human Properdin quantitates human Properdin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Properdin.
This gene encodes a plasma glycoprotein that positively regulates the alternative complement pathway of the innate immune system. This protein binds to many microbial surfaces and apoptotic cells and stabilizes the C3- and C5-convertase enzyme complexes in a feedback loop that ultimately leads to formation of the membrane attack complex and lysis of the target cell. Mutations in this gene result in two forms of properdin deficiency, which results in high susceptibility to meningococcal infections.Specifications
BCFG; Bf; BFD; C3 Proaccelerator; C3/C5 convertase; cfb; Cfp; Complement factor P; complement factor properdin; GBG; H2-Bf; PBF2; PFC; PFD; PROPERDIN; properdin factor, complement; Properdin P factor, complement | |
42 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
CFP | |
<10% | |
HRP | |
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL | |
Properdin | |
4 hr 45 min |
42 to 10,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
P27918 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |