missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Prominin 2 (AAI14526.1) VersaClone cDNA

Product Code. 18462230
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18462230 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18462230 Supplier R&D Systems™ Supplier No. RDC3197
  • ONLINE EXCLUSIVE PRICE
    £679.00
    £641.55 / 10µg
    Save £37.45
    5% Off
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

TRUSTED_SUSTAINABILITY

Specifications

Gene Prominin 2
Accession Number AAI14526.1
Gene ID (Entrez) 150696
Gene Alias hPROML2, MGC138714, PROM2, PROM-2, prominin 2, prominin-2, Prominin-like protein 2, prominin-related protein, PROML2
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.