missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Human Prolactin ELISA Kit

Product Code. 16891576 Shop All Proteintech Products
Change view
Click to view available options
Target Species:
Human
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Target Species unitSize
16891576 Human 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16891576 Supplier Proteintech™ Supplier No. KE0017296T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solid phase sandwich ELISA used to detect and quantify protein levels of endogenous PRL

  • Recognizes human PRL
  • PRL (Prolactin) is a pleiotropic hormone mainly synthesized by lactotrophs of anterior pituitary gland
    • Causes breasts to grow and make milk during pregnancy and after birth
    • Normally high levels for pregnant women and new mothers
    • Plays role in metabolism, regulation of immune system and pancreatic development
    • Normally during pregnancy prolactin serum levels increased to hold insulin resistance
    • Involved in regulation of glucose metabolism

Process:

  • Antibody specific for PRL pre-coated onto microwells
  • PRL protein in samples captured by the coated antibody after incubation
  • After extensive washing, another antibody of biotinylated specific for PRL is added to detect captured PRL protein
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm

Specifications

Accession Number NM_000948
Assay Range 31.25 to 2000 pg/mL
Assay Sensitivity 1.4 pg/ml
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human
Gene ID (Entrez) 5617
Host Species Human
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 4.5 hr.
UniProt ID P01236
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human Prolactin ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.