missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Progranulin/PGRN Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB2420-SP
This item is not returnable.
View return policy
Description
Progranulin/PGRN Monoclonal specifically detects Progranulin/PGRN in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Progranulin/PGRN | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P28799 | |
| GRN | |
| Mouse myeloma cell line NS0-derived recombinant human Progranulin/PGRN Thr18-Leu593 Accession # P28799 | |
| 25 μg | |
| Primary | |
| Detects recombinant human Progranulin/PGRN in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Progranulin/PGRN is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 296628 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in Tris and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Acrogranin, CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN, Proepithelin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2896 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction