missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Progestin Receptor beta (aa 1-50) Control Fragment Recombinant Protein

Product Code. 30209649
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209649

Brand: Invitrogen™ RP105324

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Progestin Receptor Beta-extra recognizes the extracellular epitope of the membrane progestin receptor. Progestin receptors with are found in pituitary, reproductive tract and most estrogen receptor-containing brain regions; these receptors are inducible by estrogen treatment. There are also progestin receptor sites in brain areas lacking estrogen receptors, such as the cerebral cortex of the rat; these receptors are not induced by estradiol treatment. Nevertheless, such receptors resemble those induced by estradiol. Another inducer of progestin receptors in brain is testosterone, which works through its conversion to estradiol via aromatization.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8TEZ7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 85315
Name Human Progestin Receptor beta (aa 1-50) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1700019B16Rik; 3110001D06Rik; C6orf33; LMPB1; Lysosomal membrane protein in brain 1; lysosomal membrane protein in brain-1; Membrane progesterone P4 receptor beta; Membrane progesterone receptor beta; Membrane progestin receptor beta; mPR beta; MPRB; mSR; Paqr8; Pmrbeta; PR; Progesterone and adipoQ receptor family member 8; Progestin and adipoQ receptor family member 8; progestin and adipoQ receptor family member VIII; progestin membrane receptor beta; putative membrane steroid receptor; RGD1311710
Common Name Progestin Receptor beta
Gene Symbol PAQR8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MTTAILERLSTLSVSGQQLRRLPKILEDGLPKMPCTVPETDVPQLFREPY
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.