missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pro MMP-9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9111-SP
This item is not returnable.
View return policy
Description
MMP-9 Monoclonal specifically detects MMP-9 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| MMP-9 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL | |
| P14780 | |
| MMP9 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-9 Ala20-Asp707 (Gln279Arg) Accession # P14780 | |
| 25 μg | |
| Primary | |
| Detects human Pro MMP-9 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human MMP-1, 2, 3, 7, 8, 10, 12, 13, 14, 16, or recombinant mouse MMP-9 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 526515 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP9, MMP-9, type V collagenase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4318 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction