missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Pro-MMP-13 Quantikine ELISA Kit

Product Code. p-4753677
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18712064

Brand: R&D Systems™ DM1300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Pro-MMP-13 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Pro-MMP-13 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human MMP-13 in cell culture supernates (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 78.00 to 5000 pg/mL
Assay Sensitivity 21.3 pg/mL
Conjugate HRP
Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant
Specificity Natural and Recombinant Human pro-MMP-13
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 4322
Gene Symbol MMP13
Quantity 1 Plate
Sample Volume Cell Culture Supernates (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym CLG 3, CLG3EC 3.4.24, collagenase 3, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.24, EC 3.4.24.35, EC 3.4.24.65, EC 3.4.24.7, MANDP1, matrix metallopeptidase 13 (collagenase 3), matrix metalloproteinase 13 (collagenase 3), Matrix metalloproteinase-13, MMP13, MMP-13
Target MMP-13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.