missing translation for 'onlineSavingsMsg'
Learn More

Human Pro HGF Activator Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18763399
Change view
Click to view available options
Quantity:
25 μg
500 μg
Unit Size:
25µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18763399 25 μg 25µg
18186151 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18763399 Supplier R&D Systems Supplier No. MAB1583SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HGF Activator Monoclonal specifically detects HGF Activator in Human samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HGF Activator
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 196817
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q04756
Gene Alias HGF activator, HGFA, Hgfac
Gene Symbols HGFAC
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Pro HGF Activator Gln36-Ser655 Accession # Q04756
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3083
Test Specificity Detects human Pro HGF Activator in direct ELISAs and Western blots. It recognizes only the propeptide region of HGFA precursor, not active HGFA. In Western blots, 100% cross-reactivity with recombinant mouse Pro HGFA is observed and less than 5% cross-reactivity with recombinant human (rh) Factor X and rhThrombin is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.