Learn More
Human Pro HGF Activator Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB1583
 View more versions of this product
Product Code. 18186151
  • £452.00 / 500µg
Description
HGF Activator Monoclonal specifically detects HGF Activator in Human samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HGF Activator
Monoclonal
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Q04756
HGFAC
Mouse myeloma cell line NS0-derived recombinant human Pro HGF Activator Gln36-Ser655 Accession # Q04756
500 μg
Primary
Detects human Pro HGF Activator in direct ELISAs and Western blots. It recognizes only the propeptide region of HGFA precursor, not active HGFA. In Western blots, 100% cross-reactivity with recombinant mouse Pro HGFA is observed and less than 5% cross-reactivity with recombinant human (rh) Factor X and rhThrombin is observed.
Human
Purified
Western Blot, Immunoprecipitation
196817
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
HGF activator, HGFA, Hgfac
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3083
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b