missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Pro-Gastrin-releasing Peptide DuoSet ELISA
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 5
Description
Includes
Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
The Human Pro-Gastrin-releasing Peptide DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Gastrin-releasing Peptide/Bombesin/Neuromedin C.
Specifications
Specifications
| Conjugate | Biotin |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction