missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pro-EGF Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF4289
This item is not returnable.
View return policy
Description
EGF Polyclonal specifically detects EGF in Human samples. It is validated for Western Blot.
Specifications
| EGF | |
| Polyclonal | |
| Unconjugated | |
| CAA28240 | |
| EGF | |
| Mouse myeloma cell line NS0-derived recombinant human Pro-EGF Ser21-Arg1023 Accession # CAA28240 | |
| 100 μg | |
| Primary | |
| Detects human Pro-EGF in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse Pro-EGF is observed and less than 1% cross-reactivity with mature recombinant human EGF is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG, Urogastrone | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1950 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction