missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pro Collagen I alpha 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6220-100
This item is not returnable.
View return policy
Description
Collagen I alpha 1 Monoclonal antibody specifically detects Collagen I alpha 1 in Human samples. It is validated for Immunocytochemistry.
Specifications
| Collagen I alpha 1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Alpha-1 type I collagen, COL1A1, COL-IA1, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, Collagen Type I Alpha 1 Chain, collagen, type I, alpha 1, Collagen1, Collagen-1, EDSARTH1, OI4, pro-alpha-1 collagen type 1, Type I Procollagen Alpha 1 Chain | |
| Chinese hamster ovary cell line CHO-derived recombinant human Collagen I, Gln23-Lys277, Gly1094-Leu1464, Accession # P02452 | |
| 100 μg | |
| Primary | |
| Detects human Pro-Collagen I alpha 1 in direct ELISAs. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 816161 | |
| Immunocytochemistry 8-25 ug/mL | |
| P02452 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1277 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction