missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Pro-Cathepsin B ELISA Kit

Product Code. 17154683
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17154683 96 Tests Pack of 96
17165159 5 x 96 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17154683 Supplier Invitrogen™ Supplier No. EH382RB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human Pro-Cathepsin B quantitates human Pro-Cathepsin B in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Pro-Cathepsin B.

Pro-Cathepsin B is the inactive precursor of Cathepsin B, a lysosomal cysteine protease that plays a significant role in both physiological and pathological processes. Synthesized in the endoplasmic reticulum, pro-Cathepsin B undergoes maturation in the acidic environment of the lysosome, where it is activated. Structurally, Cathepsin B consists of a heavy chain and a light chain forming a bi-lobal enzyme with a critical region known as theoccluding loop, which dictates its substrate specificity and activity. Functionally, Cathepsin B is involved in intracellular protein turnover, aiding in the degradation of proteins within endosomes and lysosomes. It also has crucial roles in extracellular matrix remodeling, facilitating processes such as cell migration, invasion, and angiogenesis, particularly in the context of cancer. Dysregulation of Cathepsin B activity is linked to various diseases, including tumor metastasis, neurodegenerative disorders, and inflammatory conditions, making it an important target for therapeutic intervention.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 61 to 15 ng/mL
Assay Sensitivity 61 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 1508
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Pro-Cathepsin B
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.