missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pro-BNP Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3604-SP
This item is not returnable.
View return policy
Description
BNP Monoclonal specifically detects BNP in Human samples. It is validated for Western Blot.
Specifications
| BNP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P16860 | |
| NPPB | |
| E. coli-derived recombinant human Pro-BNP His27-His134 Accession # P16860 | |
| 25 μg | |
| Primary | |
| Detects human Pro-BNP in direct ELISAs and Western blots. No cross-reactivity with recombinant human BNP (aa 103-134) is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 370104 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BNF, BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein, NPPB | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4879 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction