Learn More
Human Pro-BNP Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand:  R&D Systems MAB36041
Product Code. 18103421
  • £390.00 / 100µg
Description
BNP Monoclonal specifically detects BNP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BNP
Monoclonal
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL
P16860
NPPB
E. coli-derived recombinant human BNP His27-His134 Accession # P16860
100 μg
Primary
Detects human Pro-BNP in direct ELISAs. Does not detect BNP-32 (aa 103-134).
Human
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry
370124
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
BNF, BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein, NPPB
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
4879
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b