missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PRMT4 (aa 495-585) Control Fragment Recombinant Protein

Product Code. 30198731
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30198731

Brand: Invitrogen™ RP109184

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Protein arginine N-methyltransferases, such as CARM1, catalyze the transfer of a methyl group from S-adenosyl-L-methionine to the side chain nitrogens of arginine residues within proteins to form methylated arginine derivatives and S-adenosyl-L-homocysteine. Protein arginine methylation has been implicated in DNA packaging, transcription regulation, signal transduction, mRNA stability, metabolism of nascent pre-RNA, and transcriptional activation. Tissue specificity: Overexpressed in prostate adenocarcinomas and high-grade prostatic intraepithelial neoplasia.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q86X55
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10498
Name Human PRMT4 (aa 495-585) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Carm1; coactivator associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1 variant 5; histone-arginine methyltransferase CARM1; PRMT4; protein arginine methyltransferase; protein arginine N-methyltransferase 4
Common Name PRMT4
Gene Symbol Carm1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NMWNTGSTYNLSSGMAVAGMPTAYDLSSVIASGSSVGHNNLIPLGSSGAQGSGGGSTSAHYAVNSQFTMGGPAISMASPMSIPTNTMHYGS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.