missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PRL/PTP4A Pan Specific Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
£147.00 - £343.00
Specifications
| Antigen | PRL/PTP4A |
|---|---|
| Clone | 334407 |
| Concentration | LYOPH |
| Dilution | Immunohistochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18776508
![]() |
R&D Systems
MAB32191-SP |
25 μg |
£147.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18171171
|
R&D Systems
MAB32191 |
100 μg |
£343.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRL/PTP4A Monoclonal specifically detects PRL/PTP4A in Human samples. It is validated for Immunohistochemistry.Specifications
| PRL/PTP4A | |
| LYOPH | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| O75365 | |
| E. coli-derived recombinant human PRL-3 Ala2-Met173 Accession # O75365 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human PRL-2 and human PRL-3 with equal affinities in direct ELISAs. In direct ELISAs, this antibody shows 10% cross-reactivity with recombinant human PRL-1. |
| 334407 | |
| Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 7803 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title