Learn More
Abnova™ Human PRKCI (NM_002740, 1 a.a. - 587 a.a.) Full-length Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Brand: Abnova™ P4760.100ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a member of the protein kinase C (PKC) family of serine/threonine protein kinases. The PKC family comprises at least eight members, which are differentially expressed and are involved in a wide variety of cellular processes. This protein kinase is calcium-independent and phospholipid-dependent. It is not activated by phorbolesters or diacylglycerol. This kinase can be recruited to vesicle tubular clusters (VTCs) by direct interaction with the small GTPase RAB2, where this kinase phosphorylates glyceraldehyde-3-phosphate dehydrogenase (GAPD/GAPDH) and plays a role in microtubule dynamics in the early secretory pathway. This kinase is found to be necessary for BCL-ABL-mediated resistance to drug-induced apoptosis and therefore protects leukemia cells against drug-induced apoptosis. There is a single exon pseudogene mapped on chromosome X. [provided by RefSeq]
Specifications
NM_002740 | |
Functional Study, SDS-PAGE | |
5584 | |
PRKCI (Human) Recombinant Protein | |
One-step affinity purification using GSH agarose | |
100 ug | |
RUO | |
PRKCI | |
106pmol/μg x min | |
Recombinant | |
Insect cell (Sf9) expression system |
0.369 ug/uL | |
Liquid | |
98.027kDa | |
Insect cell (Sf9) expression system | |
2 ug/lane SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing | |
DXS1179E/MGC26534/PKCI/nPKC-iota | |
PRKCI | |
Baculovirus | |
GST-His | |
Liquid |