missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate MMP-9 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems MAB936-100
This item is not returnable.
View return policy
Description
MMP-9 Monoclonal specifically detects MMP-9 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| MMP-9 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP9, MMP-9, type V collagenase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4318 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, ELISA | |
| 36020 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 3-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P16035 | |
| MMP9 | |
| Chinese hamster ovary cell line CHO-derived recombinant human MMP-9 | |
| 100 μg | |
| Primary | |
| Detects human MMP-9 in Western blots. Does not react with the C-terminal truncated form (65 kDa) of recombinant human (rh) MMP-9. In Western blots, approximately 5% cross-reactivity with rhMMP-2 and no cross-reactivity with rhMMP-1 or rhMMP-3 is observed. | |
| Human, Primate | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction