missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate MMP-7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9072-500
This item is not returnable.
View return policy
Description
MMP-7 Monoclonal specifically detects MMP-7 in Human, Primate samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| MMP-7 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.24, EC 3.4.24.23, matrilysin, Matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4316 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 111439 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| NP_002414 | |
| MMP7 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-7 Leu18-Lys267 (Ala230del) Accession # NP_002414 | |
| 500 μg | |
| Primary | |
| Detects human and primate MMP-7 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) MMP-1, 2, 3, 8, 9, 10, 13, recombinant mouse MMP-7, 9, rhTIMP-1, 2, 3, or 4 is observed. | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction