missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate IL-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 66 publications
£187.00 - £1905.00
Specifications
| Antigen | IL-6 |
|---|---|
| Clone | 6708 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 0.05-0.15 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Neutralization, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18720387
![]() |
R&D Systems
MAB206-SP |
25 μg |
£187.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18130481
|
R&D Systems
MAB206-100 |
100 μg |
£471.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18186161
![]() |
R&D Systems
MAB206-500 |
500 μg |
£1905.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-6 Monoclonal specifically detects IL-6 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| IL-6 | |
| LYOPH | |
| Western Blot, Immunohistochemistry, Neutralization, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
| 3569 | |
| E. coli-derived recombinant human IL-6 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and primate IL-6 in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse (rm) IL-6, rhOSM, rhLIF, rhIL-11, rhgp130, or rhCNTF. |
| 6708 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 0.05-0.15 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human, Primate | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
| IL6 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Store the unopened product at -20°C to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title