missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate IgG Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£177.00 - £449.00
Specifications
| Antigen | IgG |
|---|---|
| Clone | 1268B |
| Dilution | Western Blot 0.05 ug/mL, Simple Western 2 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18758568
![]() |
R&D Systems
MAB1101-SP |
25 μg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18730018
![]() |
R&D Systems
MAB1101-100 |
100 μg |
£449.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgG Monoclonal specifically detects IgG in Human, Primate samples. It is validated for Western Blot, Simple Western.Specifications
| IgG | |
| Western Blot 0.05 ug/mL, Simple Western 2 ug/mL | |
| Monoclonal | |
| Rabbit | |
| Human, Primate | |
| Immunoglobulin G, ImmunoglobulinG | |
| Human IgG | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and primate IgG in Western blots. |
| 1268B | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 3500 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title