missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate EGF Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB636-500
This item is not returnable.
View return policy
Description
EGF Monoclonal specifically detects EGF in Human, Primate samples. It is validated for Immunohistochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| EGF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EGF | |
| E. coli-derived recombinant human EGF | |
| 500 μg | |
| Primary | |
| Detects human and primate EGF in ELISAs. In sandwich ELISAs, no cross-reactivity or interference is observed with recombinant mouse (rm) EGF, bovine FGF acidic, bovine FGF basic, recombinant human (rh) G-CSF, rhGM-CSF, rmGM-CSF, rhLIF, human PDGF, porcine PDGF, rhTGF-alpha , hTGF-beta 1, pTGF-beta 1, rhTGF-beta 1, porcine TGF-beta 1.2, porcine TGF-beta 2, rhTNF-alpha , or rhTNF-beta . | |
| Human, Primate | |
| Supernatant |
| Immunohistochemistry, ELISA | |
| 10827 | |
| Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG, Urogastrone | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1950 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction