missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pref-1/DLK1/FA1 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
£238.00 - £450.00
Specifications
| Antigen | Pref-1/DLK1/FA1 |
|---|---|
| Clone | 211309 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18798047
![]() |
R&D Systems
FAB1144G-025 |
25 Tests |
£238.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18795037
![]() |
R&D Systems
FAB1144G-100 |
100 Tests |
£450.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pref-1/DLK1/FA1 Monoclonal specifically detects Pref-1/DLK1/FA1 in Human samples. It is validated for Flow Cytometry.Specifications
| Pref-1/DLK1/FA1 | |
| Flow Cytometry 5 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| delta-like 1 homolog (Drosophila), DLK, DLK1, DLK-1, FA1, FA1fetal antigen 1, pG2, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG | |
| DLK1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human Pref-1/DLK1/FA1 in direct ELISAs and Western blots. Shows approximately 50% cross-reactivity with recombinant mouse Pref-1/DLK1/FA1. |
| 211309 | |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Mouse | |
| Human | |
| P80370 | |
| 8788 | |
| Mouse myeloma cell line NS0-derived recombinant human Pref-1/DLK1/FA1 long isoform Ala24-Pro297 (with Arg248Pro and Lys295Ser substitutions) Accession # P80370 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title