missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PPP1R3D (aa 112-187) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59450 (PA5-59450. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Seems to act as a glycogen-targeting subunit for PP1. PP1 is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis.
Specifications
Specifications
| Accession Number | O95685 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 5509 |
| Name | Human PPP1R3D (aa 112-187) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2610024M03Rik; Gm359; PP1 subunit R6; PPP1R3D; PPP1R6; protein phosphatase 1 regulatory subunit 3 D; Protein phosphatase 1 regulatory subunit 6; protein phosphatase 1, regulatory (inhibitor) subunit 3 D; protein phosphatase 1, regulatory subunit 3 D; protein phosphatase 1, regulatory subunit 6, spinophilin; protein phosphatase 1-binding subunit R6 |
| Common Name | PPP1R3D |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction