missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PPIG Control Fragment Recombinant Protein

Product Code. 30201043
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201043

Brand: Invitrogen™ RP103943

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84346 (PA5-84346. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Structural Maintenance of Chromosomes (SMC) family proteins play critical roles in various nuclear events that require structural changes of chromosomes, including mitotic chromosome organization, DNA recombination and repair and global transcriptional repression. The chromo proteins are conserved in eukaryotes and lead to mitotic chromosome segregation defects. This suggests a critical function of SMC family proteins in mitotic chromosome dynamics. SMC1 and SMC3 form a heterodimeric complex required for metaphase progression in mitotic cells. Specifically this SMC1/SMC3 complex is responsible for sister chromatid cohesion during metaphase. Mutations in the gene can lead to Cornelia de Lange syndrome 2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13427
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9360
Name Human PPIG Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AU019516; AU022200; B230312B02Rik; CARS-cyclophilin; CARS-Cyp; CASP10; Clk-associating RS-cyclophilin; Cyclophilin G; CYP; Matrin cyclophilin; matrin cyclophilin (matrin-cyp); matrin-cyp; MGC133241; OTTHUMP00000204761; OTTHUMP00000204762; Peptidyl-prolyl cis-trans isomerase G; peptidylprolyl isomerase G; peptidyl-prolyl isomerase G; peptidylprolyl isomerase G (cyclophilin G); peptidyl-prolyl isomerase G (cyclophilin G); PPIase G; PPIG; Rotamase G; SCAF10; SR-cyclophilin; SRCyp; SR-cyp; SR-related CTD-associated factor 10
Common Name PPIG
Gene Symbol PPIG
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QRMRVSSGERWIKGDKSELNEIKENQRSPVRVKERKITDHRNVSESPNRKNEKEKKVKDHKSNSKERDIRRNSEKEDKYKNKVKKRAKSKS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.