missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PP4 Catalytic Subunit Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB5074
This item is not returnable.
View return policy
Description
PP4/PPP4C Monoclonal specifically detects PP4/PPP4C in Human samples. It is validated for Western Blot.
Specifications
| PP4/PPP4C | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P60510 | |
| PPP4C | |
| E. coli-derived recombinant human PP4 Catalytic Subunit Met1-Leu307 Accession # P60510 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat PP4 Catalytic Subunit in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 501517 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.1.3.16, PP4C, PP4protein phosphatase X, catalytic subunit, PPH3, PPP4C, PPX, PP-X, PPXPPH3, protein phosphatase 4 (formerly X), catalytic subunit, protein phosphatase 4, catalytic subunit, Protein phosphatase X | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5531 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction