missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PP4 Catalytic Subunit Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB5074-SP
This item is not returnable.
View return policy
Description
PP4/PPP4C Monoclonal specifically detects PP4/PPP4C in Human samples. It is validated for Western Blot.
Specifications
| PP4/PPP4C | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P60510 | |
| PPP4C | |
| E. coli-derived recombinant human PP4 Catalytic Subunit Met1-Leu307 Accession # P60510 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat PP4 Catalytic Subunit in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 501517 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.1.3.16, PP4C, PP4protein phosphatase X, catalytic subunit, PPH3, PPP4C, PPX, PP-X, PPXPPH3, protein phosphatase 4 (formerly X), catalytic subunit, protein phosphatase 4, catalytic subunit, Protein phosphatase X | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5531 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction