missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PP14 ELISA Kit
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
Description
Human PP14 quantitates human PP14 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PP14.
Human placental protein 14 (PP14; also known as glycodelin and progesterone-associated endometrial protein) is a protein of the lipocalin structural superfamily. PP14 is the most abundant product of the secretory endometrium, and has been proposed as a biochemical marker of endometrial function in women undergoing in vitro fertilization treatment.
Specifications
Specifications
| Accession Number | alpha uterine protein; GD; GdA; GdF; GdS; Glycodelin; glycodelin-A; glycodelin-F; glycodelin-S; PAEG; PAEP; PEG; PEP; Placental protein 14; PP14; PP14 protein (placental protein 14); pregnancy-associated endometrial alpha-2 globulin; pregnancy-associated endometrial alpha-2-globulin; progestagen associated endometrial protein; Progestagen-associated endometrial protein; progestagen-associated endometrial protein (placental protein 14, pregnancy-associated endometrial a; Progesterone-associated endometrial protein; ZIF-1; Zona-binding inhibitory factor-1) |
| Assay Range | 2.5 to 600 ng/mL |
| Assay Sensitivity | 2.5 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 5047 |
| Gene Symbol | PAEP |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction