missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human POT1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5299-SP
This item is not returnable.
View return policy
Description
POT1 Monoclonal specifically detects POT1 in Human samples. It is validated for Western Blot.
Specifications
| POT1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q9NUX5 | |
| POT1 | |
| E. coli-derived recombinant human POT1 Met159-Pro316 Accession # Q9NUX5 | |
| 25 μg | |
| Primary | |
| Detects human POT1 in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot | |
| 688923 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp586D211, hPot1, POT1 protection of telomeres 1 homolog, POT1-like telomere end-binding protein, protection of telomeres 1 homolog (S. pombe), protection of telomeres protein 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 25913 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction