missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Porimin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3010-SP
This item is not returnable.
View return policy
Description
Porimin Monoclonal specifically detects Porimin in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| Porimin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| AAG21694 | |
| TMEM123 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Porimin His27-Ser149 Accession # AAG21694 | |
| 25 μg | |
| Primary | |
| Detects human Porimin in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 297617 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KCT3, KCT-3, KCT3keratinocytes associated transmembrane protein 3, Keratinocytes-associated transmembrane protein 3, PORIMINporimin, pro oncosis receptor inducing membrane injury, Pro-oncosis receptor inducing membrane injury, serine/threonine-rich receptor, TMEM123, transmembrane protein 123PORMIN | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 114908 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction