missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Porimin Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB3010V-100UG
This item is not returnable.
View return policy
Description
Porimin Monoclonal specifically detects Porimin in Human samples. It is validated for Flow Cytometry.
Specifications
| Porimin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| AAG21694 | |
| TMEM123 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Porimin His27-Ser149 Accession # AAG21694 | |
| RUO | |
| 114908 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 297617 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| KCT3, KCT-3, KCT3keratinocytes associated transmembrane protein 3, Keratinocytes-associated transmembrane protein 3, PORIMINporimin, pro oncosis receptor inducing membrane injury, Pro-oncosis receptor inducing membrane injury, serine/threonine-rich receptor, TMEM123, transmembrane protein 123PORMIN | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Porimin in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction