missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human POR/Cytochrome P450 Reductase Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6340-SP
This item is not returnable.
View return policy
Description
POR/Cytochrome P450 Reductase Monoclonal specifically detects POR/Cytochrome P450 Reductase in Human samples. It is validated for Western Blot.
Specifications
| POR/Cytochrome P450 Reductase | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P16435 | |
| POR | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human POR/Cytochrome P450 Reductase Arg45-Ser677 Accession # P16435 | |
| 25 μg | |
| Primary | |
| Detects human POR/Cytochrome P450 Reductase in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot | |
| 739613 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CPR, CYPOR, CYPORFLJ26468, Cytochrome P450 Reductase, DKFZp686G04235, EC 1.6.2.4, NADPH--cytochrome P450 reductase, NADPH-dependent cytochrome P450 reductase, P450 (cytochrome) oxidoreductase, P450R | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5447 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction