missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PON1 Recombinant Protein
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
This product contains purified recombinant human Paraoxonase 1 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
The paraoxonase (PON) gene-family in humans has three members, PON1, PON2, and PON3, aligned next to each other on the long arm of chromosome 7q21.3-22.1. PON1 of these is synthesized primarily in the liver and a portion is secreted into the plasma, where it is associated with high density lipoproteins (HDL). And It hydrolyzes the active metabolites of several other organophosphorus insecticides (e.g., chlorpyrifos oxon, diazoxon), as well as nerve agents such as sarin, soman and VX (lethal nerve agent). The PON1 polymorphism may be responsible for neurodegeneration and considered to be an independent risk factor for Parkinson's disease.
Specifications
Specifications
| Accession Number | P27169 |
| Concentration | 0.69 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 20 mM tris HCl with 50% glycerol and no preservative; pH 8 |
| Gene ID (Entrez) | 5444 |
| Molecular Weight (g/mol) | 44.26 kDa |
| Name | Human PON1 |
| Quantity | 1 mg |
| Source | E. coli |
| Regulatory Status | RUO |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido