missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PON1 ELISA Kit
Description
Human PON1 quantitates human PON1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PON1.
The paraoxonase (PON) gene-family in humans has three members, PON1, PON2, and PON3, aligned next to each other on the long arm of chromosome 7q21.3-22.1. PON1 of these is synthesized primarily in the liver and a portion is secreted into the plasma, where it is associated with high density lipoproteins (HDL). And It hydrolyzes the active metabolites of several other organophosphorus insecticides (e.g., chlorpyrifos oxon, diazoxon), as well as nerve agents such as sarin, soman and VX (lethal nerve agent). The PON1 polymorphism may be responsible for neurodegeneration and considered to be an independent risk factor for Parkinson's disease.
Specifications
Specifications
| Accession Number | A esterase 1; A-esterase 1; aromatic esterase 1; arylesterase 1; arylesterase B-type; EC 3.1.1.2; EC 3.1.1.81; EC 3.1.8.1; ESA; ESAesterase A; esterase A; K-45; MVCD5; paraoxonase 1; paraoxonase B-type; Pon; PON 1; PON1; serum aryldiakylphosphatase; serum aryldialkylphosphatase 1; Serum paraoxonase/arylesterase 1 |
| Assay Range | 0.2 to 50 ng/mL |
| Assay Sensitivity | 0.2 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 5444 |
| Gene Symbol | PON1 |
| Interassay CV | <12% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?