missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Polypeptide GalNAc Transferase 1/GALNT1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7140-SP
This item is not returnable.
View return policy
Description
Polypeptide GalNac Transferase 1/GALNT1 Polyclonal specifically detects Polypeptide GalNac Transferase 1/GALNT1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Polypeptide GalNac Transferase 1/GALNT1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.4.1.41, GalNAc transferase 1, GALNAC-T1, Polypeptide GalNAc transferase 1, polypeptide N-acetylgalactosaminyltransferase 1, pp-GaNTase 1, Protein-UDP acetylgalactosaminyltransferase 1, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 1, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 1 (GalNAc-T1) | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2589 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q10472 | |
| GALNT1 | |
| Mouse myeloma cell line NS0-derived recombinant human Polypeptide GalNac Transferase 1/GALNT1 Gly41-Phe559 Accession # Q10472 | |
| 25 μg | |
| Primary | |
| Detects human Polypeptide GalNac Transferase 1/GALNT1 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) GALNT2 and rhGALNT3 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction