missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human POLR3C Full-length ORF (NP_006459.3, 1 a.a. - 534 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010623-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16165442

  • £280.00 / 10µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MTQAEIKLCSLLLQEHFGEIVEKIGVHLIRTGSQPLRVIAHDTGTSLDQVKKALCVLVQHNLVSYQVHKRGVVEYEAQCSRVLRMLRYPRYIYTTKTLYSDTGELIVEELLLNGKLTMSAVVKKVADRLTETMEDGKTMDYAEVSNTFVRLADTHFVQRCPSVPTTENSDPGPPPPAPTLVINEKDMYLVPKLSLIGKGKRRRSSDEDAAGEPKAKRPKYTTDNKEPIPDDGIYWQANLDRFHQHFRDQAIVSAVANRMDQTSSEIVRTMLRMSEITTSSSAPFTQPLSSNEIFRSLPVGYNISKQVLDQYLTLLADDPLEFVGKSGDSGGGMYVINLHKALASLATATLESVVQERFGSRCARIFRLVLQKKHIEQKQVEDFAMIPAKEAKDMLYKMLSENFMSLQEIPKTPDHAPSRTFYLYTVNILSAARMLLHRCYKSIANLIERRQFETKENKRLLEKSQRVEAIIASMQATGAEEAQLQEIEEMITAPERQQLETLKRNVNKLDASEIQVDETIFLLESYIECTMKRQ
Specifications

Specifications

NP_006459.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
87kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RPC3/RPC62
POLR3C
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10623
POLR3C (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTQAEIKLCSLLLQEHFGEIVEKIGVHLIRTGSQPLRVIAHDTGTSLDQVKKALCVLVQHNLVSYQVHKRGVVEYEAQCSRVLRMLRYPRYIYTTKTLYSDTGELIVEELLLNGKLTMSAVVKKVADRLTETMEDGKTMDYAEVSNTFVRLADTHFVQRCPSVPTTENSDPGPPPPAPTLVINEKDMYLVPKLSLIGKGKRRRSSDEDAAGEPKAKRPKYTTDNKEPIPDDGIYWQANLDRFHQHFRDQAIVSAVANRMDQTSSEIVRTMLRMSEITTSSSAPFTQPLSSNEIFRSLPVGYNISKQVLDQYLTLLADDPLEFVGKSGDSGGGMYVINLHKALASLATATLESVVQERFGSRCARIFRLVLQKKHIEQKQVEDFAMIPAKEAKDMLYKMLSENFMSLQEIPKTPDHAPSRTFYLYTVNILSAARMLLHRCYKSIANLIERRQFETKENKRLLEKSQRVEAIIASMQATGAEEAQLQEIEEMITAPERQQLETLKRNVNKLDASEIQVDETIFLLESYIECTMKRQ
RUO
POLR3C
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers