missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human POLR3B (aa 191-282) Control Fragment Recombinant Protein

Product Code. 30181343
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30181343

Brand: Invitrogen™ RP97632

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57671 (PA5-57671. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

POLR3B (RNA Polymerase III Subunit B) is a Protein Coding gene. Diseases associated with POLR3B include Leukodystrophy, Hypomyelinating, 8, With Or Without Oligodontia And/Or Hypogonadotropic Hypogonadism and Cerebellar Hypoplasia With Endosteal Sclerosis. Among its related pathways are RNA Polymerase II Transcription Initiation And Promoter Clearance and ATP/ITP metabolism. Gene Ontology (GO) annotations related to this gene include DNA-directed 5'-3' RNA polymerase activity and ribonucleoside binding. An important paralog of this gene is POLR2B.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NW08
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55703
Name Human POLR3B (aa 191-282) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2700078H01Rik; A330032P03Rik; C128; C85372; DNA-directed RNA polymerase III 127.6 kDa polypeptide; DNA-directed RNA polymerase III B; DNA-directed RNA polymerase III subunit B; DNA-directed RNA polymerase III subunit RPC2; FLJ10388; HLD8; INMAP; interphase nucleus and mitotic apparatus associated protein; POLR3B; polymerase (RNA) III (DNA directed) polypeptide B; polymerase (RNA) III subunit B; RGD1565311; RNA polymerase III subunit B; RNA polymerase III subunit C2; RNA polymerase III subunit RPC2 homolog; RPC2; si:dkey-103i16.3; slim jim; slj; wu:fc20h06
Common Name POLR3B
Gene Symbol POLR3B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VEADRKGAVGASVTSSTHEKKSRTNMAVKQGRFYLRHNTLSEDIPIVIIFKAMGVESDQEIVQMIGTEEHVMAAFGPSLEECQKAQIFTQMQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.