missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human POLR1C (aa 96-195) Control Fragment Recombinant Protein

Product Code. 30207740
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30207740

Brand: Invitrogen™ RP94949

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111008 (PA5-111008. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

RNA polymerases transcribe nuclear genes for ribosomal RNA, thus representing ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. Different subunits of the Pol I transcription machinery are targets of various physiological stimuli, which suggests that multiple signaling pathways are involved in carrying out Pol I transcription. RPA16, RPA40 and RPA135 are subunits of Pol I that associate with each other at an early stage of RNA Pol I assembly. RPA40 is essential for the function and integrity of the complex and is also an essential subunit of RNA polymerase III (Pol III).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15160
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9533
Name Human POLR1C (aa 96-195) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 40 kDa; AA409007; AA959927; Ac2-127; AC40; AL024089; DNA-directed RNA polymerase I subunit C; DNA-directed RNA polymerases I and III 40 kDa polypeptide; DNA-directed RNA polymerases I and III subuni RPAC1-like protein; DNA-directed RNA polymerases I and III subunit RPAC1; HLD11; Polr1c; POLR1E; polymerase (RNA) I polypeptide C; polymerase (RNA) I polypeptide C, 30 kDa; polymerase (RNA) I subunit C; RNA polymerase 1-1; RNA polymerase 1-1 (40 kDa subunit); RNA polymerase I 140 kDa subunit; RNA polymerase I and III subunit C; RNA polymerase I subunit; RNA polymerase I subunit C; RNA polymerase subunit I; RNA polymerases I and III subunit AC1; RPA39; RPA40; RPA5; RPAC1; RPC40; Rpo1-1; TCS3; Unknown (protein for MGC:134521); zgc:65973
Common Name POLR1C
Gene Symbol POLR1C
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NNTSIVQDEILAHRLGLIPIHADPRLFEYRNQGDEEGTEIDTLQFRLQVRCTRNPHAAKDSSDPNELYVNHKVYTRHMTWIPLGNQADLFPEGTIRPVHD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.