missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human POLG2 (aa 1-94) Control Fragment Recombinant Protein

Product Code. 30198669
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30198669

Brand: Invitrogen™ RP109904

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (37%), Rat (37%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-145191 (PA5-145191. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes the processivity subunit of the mitochondrial DNA polymerase gamma. The encoded protein forms a heterotrimer containing one catalytic subunit and two processivity subunits. This protein enhances DNA binding and promotes processive DNA synthesis. Mutations in this gene result in autosomal dominant progressive external ophthalmoplegia with mitochondrial DNA deletions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UHN1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 11232
Name Human POLG2 (aa 1-94) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA409516; DNA polymerase gamma; DNA polymerase gamma 2, accessory subunit; DNA polymerase gamma accessory 55 kDa subunit; DNA polymerase gamma subunit 1 (Mitochondrial DNA polymerase catalytic subunit) (PolG-alpha); DNA polymerase gamma, catalytic subunit; DNA polymerase subunit gamma-1; DNA polymerase subunit gamma-2, mitochondrial; DNA-directed DNA polymerase gamma; HP55; MDP1; Mip1; MIRAS; Mitochondrial DNA polymerase accessory subunit; mitochondrial DNA polymerase catalytic subunit; mitochondrial DNA polymerase gamma; mitochondrial DNA polymerase gamma catalytic subunit; mitochondrial DNA polymerase subunit gamma-2; mitochondrial DNA polymerase, accessory subunit; mitochondrial DNA polymerase-gamma; mitochondrial polymerase gamma; mitochondrial polymerase gamma 2; MTDPS4A; MTDPS4B; MTPOLB; p55; PEO; PEOA4; Pol gamma; POLB; POLG; POLG1; Polg2; POLGA; PolG-alpha; POLGB; polG-beta; polymerase (DNA directed), gamma; polymerase (DNA directed), gamma 2, accessory subunit; polymerase (DNA) gamma 2, accessory subunit; polymerase (DNA) gamma, catalytic subunit; polymerase, gamma; polymerase, gamma 2; SANDO; SCAE; truncated mitochondrial DNA polymerase gamma catalytic subunit
Common Name POLG2
Gene Symbol POLG2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MRSRVAVRACHKVCRCLLSGFGGRVDAGQPELLTERSSPKGGHVKSHAELEGNGEHPEAPGSGEGSEALLEICQRRHFLSGSKQQLSRDSLLSG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.