missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PNAS4 (aa 7-96) Control Fragment Recombinant Protein

Product Code. 30181639
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30181639

Brand: Invitrogen™ RP99772

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65589 (PA5-65589. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PNAS4 is a highly conserved protein that shares high homology from plants to animals. It was initially identified as a putative apoptosis-related protein in the human acute promyelocytic leukemia cell line NB4. PNAS4 is activated during the early response to DNA damage and can inhibit proliferation via apoptosis when overexpressed in some tumor cells such as U2OS, SKOV3, and A549. PNAS4 inhibits tumor cell proliferation through the following mechanisms: (i) overexpression of PNAS4 causes S phase arrest by regulating the expression of cell cycle-related proteins and (ii) PNAS4 induces apoptosis through the mitochondrial apoptosis pathway. Recent evidence has shown that PNAS4 may be involved in the genesis of some cancers and could be a potential candidate for lung cancer therapy alone or in combination with gemcitabine. At least two isoforms of PNAS4 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BSY9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51029
Name Human PNAS4 (aa 7-96) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5830417C01Rik; C1orf121; cb238; CGI-146; DESI; DESI1; desi2; DeSI-2; desumoylating isopeptidase 1; desumoylating isopeptidase 2; Deubiquitinase DESI2; Fam152a; family with sequence similarity 152, member A; fi12d12; hypothetical protein LOC445138; pnas4; PNAS-4; PPPDE peptidase domain containing 1; PPPDE peptidase domain-containing protein 1; Pppde1; Protein FAM152A; sb:cb238; si:ch211-132e22.3; wu:fi12d12; zgc:100860
Common Name PNAS4
Gene Symbol DESI2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VVLNVYDMYWMNEYTSSIGIGVFHSGIEVYGREFAYGGHPYPFSGIFEISPGNASELGETFKFKEAVVLGSTDFLEDDIEKIVEELGKEY
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.