missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PMF1 (aa 3-70) Control Fragment Recombinant Protein

Product Code. 30204292
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30204292

Brand: Invitrogen™ RP104646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (57%), Rat (57%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65125 (PA5-65125. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PMF1 is part of the MIS12 complex which is required for normal chromosome alignment and segregation and kinetochore formation during mitosis. It may act as a cotranscription partner of NFE2L2 involved in regulation of polyamine-induced transcription of SSAT.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6P1K2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 11243
Name Human PMF1 (aa 3-70) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2600009M07Rik; AL033286; AW060657; Est1p-like protein B (EST1B); Pmf1; PMF-1; polyamine modulated factor 1; polyamine-modulated factor 1; RP11-54H19.4; Unknown (protein for MGC:134238)
Common Name PMF1
Gene Symbol PMF1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EASSANLGSGCEEKRHEGSSSESVPPGTTISRVKLLDTMVDTFLQKLVAAGSYQRFTDCYKCFYQLQP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.