missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PLXNB3 (aa 1031-1137) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (78%), Rat (78%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61638 (PA5-61638. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The protein encoded by this gene is a member of the plexin family. It functions as a receptor for semaphorin 5A, and plays a role in axon guidance, invasive growth and cell migration. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Accession Number | Q9ULL4 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 5365 |
| Name | Human PLXNB3 (aa 1031-1137) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AI451018; Ensembl:ENSRNOP00000053235}; FLJ76953; KIAA1206; PLEXB3; plexin 6; plexin B3; plexin-6; Plexin-B3; plexin-B3 {ECO:0000303; PLEXR; Plxn6; PLXNB3; plxnb3 {ECO:0000312; Protein Plxnb3; PubMed:20696765}; RGD1560615 |
| Common Name | PLXNB3 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction