missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Pluripotent Stem Cell Naïve State qPCR Array
Description
- Characterize your hPSCs by accurately measuring their gene expression profile to determine their status along the spectrum from naïve to primed pluripotency
- The Human Pluripotent Stem Cell (hPSC) Naïve State qPCR Array has validated primers and probes to detect 90 genes preselected for their demonstrated differential expression in naïve state and primed hPSCs and their derivatives, as well as 6 endogenous control genes
- TBP (TATA box-binding protein) qPCR Control Template is provided as a synthetic DNA positive control
- Naïve state hPSCs are self-renewing, retain the properties of the pre-implantation blastocyst, and can differentiate to both embryonic and extra-embryonic lineages
- Primed hPSCs are also self-renewing, but they are restricted to differentiation along the three embryonic germ layers
Specifications
Specifications
| Description | Pluripotent Stem Cells |
| Format | 96-well Plate Type 4 |
| For Use With (Application) | Real Time PCR (qPCR) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?